RIE156322353 Rieke Metals, Inc.

2-(3-Methylbutyryl)oxazole

  • MDL number MFCD07699325
  • Cas Number : 898759-16-5
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-12
Availability details for
Quantity
  • Availability
  • Add to Cart