RIE156322354 Rieke Metals, Inc.

2-(3,3-Dimethylbutyryl)oxazole

  • MDL number MFCD07699326
  • Cas Number : 898759-18-7
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-13
Availability details for
Quantity
  • Availability
  • Add to Cart