RIE156322355 Rieke Metals, Inc.

2-(2-Methylvaleryl)oxazole

  • MDL number MFCD07699328
  • Cas Number : 898759-23-4
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-1-(1,3-oxazol-2-yl)-1-pentanone
Inchi Code 1S/C9H13NO2/c1-3-4-7(2)8(11)9-10-5-6-12-9/h5-7H,3-4H2,1-2H3
Inchi Key SDWUEGRLDHTQOQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5571-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-15
Availability details for
Quantity
  • Availability
  • Add to Cart