RIE156322356 Rieke Metals, Inc.

2-(3-Methylvaleryl)oxazole

  • MDL number MFCD07699329
  • Cas Number : 898759-26-7
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-16
Availability details for
Quantity
  • Availability
  • Add to Cart