RIE156322357 Rieke Metals, Inc.

2-(4-Methylvaleryl)oxazole

  • MDL number MFCD07699330
  • Cas Number : 898759-29-0
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-17
Availability details for
Quantity
  • Availability
  • Add to Cart