RIE156322360 Rieke Metals, Inc.

2-(2-Propylvaleryl)oxazole

  • MDL number MFCD07699333
  • Cas Number : 898759-38-1
  • Molecular Weight : 195.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-20
Availability details for
Quantity
  • Availability
  • Add to Cart