RIE156322362 Rieke Metals, Inc.

2-(2-Methoxybenzoyl)oxazole

  • MDL number MFCD07699336
  • Cas Number : 898759-44-9
  • Molecular Weight : 203.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-02
Availability details for
Quantity
  • Availability
  • Add to Cart