RIE156322366 Rieke Metals, Inc.

2-(3-Methylbenzoyl)oxazole

  • MDL number MFCD07699340
  • Cas Number : 898759-56-3
  • Molecular Weight : 187.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-06
Availability details for
Quantity
  • Availability
  • Add to Cart