RIE156322370 Rieke Metals, Inc.

2-(4-Trifluoromethylbenzoyl)oxazole

  • MDL number MFCD07699344
  • Cas Number : 898759-66-5
  • Molecular Weight : 241.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-oxazol-2-yl[4-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C11H6F3NO2/c12-11(13,14)8-3-1-7(2-4-8)9(16)10-15-5-6-17-10/h1-6H
Inchi Key IHUHMIRJRFVXFH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5572-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-10
Availability details for
Quantity
  • Availability
  • Add to Cart