RIE156322375 Rieke Metals, Inc.

2-(4-Chlorobenzoyl)oxazole

  • MDL number MFCD07699350
  • Cas Number : 898759-77-8
  • Molecular Weight : 207.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chlorophenyl)(1,3-oxazol-2-yl)methanone
Inchi Code 1S/C10H6ClNO2/c11-8-3-1-7(2-4-8)9(13)10-12-5-6-14-10/h1-6H
Inchi Key ARNJPEUSZIHOQW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5572-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-16
Availability details for
Quantity
  • Availability
  • Add to Cart