RIE156322378 Rieke Metals, Inc.

2-(4-Bromobenzoyl)oxazole

  • MDL number MFCD07699353
  • Cas Number : 698355-80-5
  • Molecular Weight : 252.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-19
Availability details for
Quantity
  • Availability
  • Add to Cart