RIE156322381 Rieke Metals, Inc.

2-(4-Iodobenzoyl)oxazole

  • MDL number MFCD07699356
  • Cas Number : 898759-87-0
  • Molecular Weight : 299.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-22
Availability details for
Quantity
  • Availability
  • Add to Cart