RIE156322387 Rieke Metals, Inc.

2-(4-Nitrobenzoyl)oxazole

  • MDL number MFCD07782996
  • Cas Number : 898759-91-6
  • Molecular Weight : 218.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-nitrophenyl)(1,3-oxazol-2-yl)methanone
Inchi Code 1S/C10H6N2O4/c13-9(10-11-5-6-16-10)7-1-3-8(4-2-7)12(14)15/h1-6H
Inchi Key SOTWEQBYCNXMLO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5572-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-31
Availability details for
Quantity
  • Availability
  • Add to Cart