RIE156322390 Rieke Metals, Inc.

2-(4-t-Butylbenzoyl)oxazole

  • MDL number MFCD07699363
  • Cas Number : 898760-03-7
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-34
Availability details for
Quantity
  • Availability
  • Add to Cart