RIE156322392 Rieke Metals, Inc.

2-(4-Propylbenzoyl)oxazole

  • MDL number MFCD07699365
  • Cas Number : 898760-07-1
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-oxazol-2-yl(4-propylphenyl)methanone
Inchi Code 1S/C13H13NO2/c1-2-3-10-4-6-11(7-5-10)12(15)13-14-8-9-16-13/h4-9H,2-3H2,1H3
Inchi Key ABLNLEMDZDCPIR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5572-35b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-35b
Availability details for
Quantity
  • Availability
  • Add to Cart