RIE156322394 Rieke Metals, Inc.

2-(4-Pentylbenzoyl)oxazole

  • MDL number MFCD07699367
  • Cas Number : 898760-11-7
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-35D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-35D
Availability details for
Quantity
  • Availability
  • Add to Cart