RIE156322395 Rieke Metals, Inc.

2-(4-Hexylbenzoyl)oxazole

  • MDL number MFCD07699368
  • Cas Number : 898760-13-9
  • Molecular Weight : 257.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-35E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-35E
Availability details for
Quantity
  • Availability
  • Add to Cart