RIE156322396 Rieke Metals, Inc.

2-(4-Heptylbenzoyl)oxazole

  • MDL number MFCD07699369
  • Cas Number : 898760-15-1
  • Molecular Weight : 271.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-35F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-35F
Availability details for
Quantity
  • Availability
  • Add to Cart