RIE156322397 Rieke Metals, Inc.

2-(4-Ethoxybenzoyl)oxazole

  • MDL number MFCD07699370
  • Cas Number : 898760-17-3
  • Molecular Weight : 217.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-ethoxyphenyl)(1,3-oxazol-2-yl)methanone
Inchi Code 1S/C12H11NO3/c1-2-15-10-5-3-9(4-6-10)11(14)12-13-7-8-16-12/h3-8H,2H2,1H3
Inchi Key ITTBPAVMPMOSSO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5572-36a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-36a
Availability details for
Quantity
  • Availability
  • Add to Cart