RIE156322398 Rieke Metals, Inc.

2-(4-Propoxybenzoyl)oxazole

  • MDL number MFCD07699371
  • Cas Number : 898760-20-8
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-36B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-36B
Availability details for
Quantity
  • Availability
  • Add to Cart