RIE156322399 Rieke Metals, Inc.

2-(4-Butoxybenzoyl)oxazole

  • MDL number MFCD07699372
  • Cas Number : 898760-23-1
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-36C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-36C
Availability details for
Quantity
  • Availability
  • Add to Cart