RIE156322400 Rieke Metals, Inc.

2-(4-Pentyloxybenzoyl)oxazole

  • MDL number MFCD07699373
  • Cas Number : 898760-26-4
  • Molecular Weight : 259.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-36D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-36D
Availability details for
Quantity
  • Availability
  • Add to Cart