RIE156322401 Rieke Metals, Inc.

2-(4-Hexyloxybenzoyl)oxazole

  • MDL number MFCD07699374
  • Cas Number : 898760-29-7
  • Molecular Weight : 273.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-36E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-36E
Availability details for
Quantity
  • Availability
  • Add to Cart