RIE156322405 Rieke Metals, Inc.

2-(2,5-Difluorobenzoyl)oxazole

  • MDL number MFCD07699378
  • Cas Number : 898760-41-3
  • Molecular Weight : 209.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-39

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-39
Availability details for
Quantity
  • Availability
  • Add to Cart