RIE156322406 Rieke Metals, Inc.

2-(2,6-Difluorobenzoyl)oxazole

  • MDL number MFCD07699379
  • Cas Number : 898760-44-6
  • Molecular Weight : 209.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,6-difluorophenyl)(1,3-oxazol-2-yl)methanone
Inchi Code 1S/C10H5F2NO2/c11-6-2-1-3-7(12)8(6)9(14)10-13-4-5-15-10/h1-5H
Inchi Key VAZIGHLZLIEDIZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5572-40

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-40
Availability details for
Quantity
  • Availability
  • Add to Cart