RIE156322408 Rieke Metals, Inc.

2-(3,5-Difluorobenzoyl)oxazole

  • MDL number MFCD07699381
  • Cas Number : 898760-50-4
  • Molecular Weight : 209.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-difluorophenyl)(1,3-oxazol-2-yl)methanone
Inchi Code 1S/C10H5F2NO2/c11-7-3-6(4-8(12)5-7)9(14)10-13-1-2-15-10/h1-5H
Inchi Key UHAJIVGLVWBQGR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5572-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-42
Availability details for
Quantity
  • Availability
  • Add to Cart