RIE156322411 Rieke Metals, Inc.

2-(2,5-Dichlorobenzoyl)oxazole

  • MDL number MFCD07699384
  • Cas Number : 898784-22-0
  • Molecular Weight : 242.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,5-dichlorophenyl)(1,3-oxazol-2-yl)methanone
Inchi Code 1S/C10H5Cl2NO2/c11-6-1-2-8(12)7(5-6)9(14)10-13-3-4-15-10/h1-5H
Inchi Key NVYOVJNELYQXPB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5572-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-45
Availability details for
Quantity
  • Availability
  • Add to Cart