RIE156322414 Rieke Metals, Inc.

2-(3,5-Dichlorobenzoyl)oxazole

  • MDL number MFCD07699387
  • Cas Number : 898784-28-6
  • Molecular Weight : 242.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-48
Availability details for
Quantity
  • Availability
  • Add to Cart