RIE156322420 Rieke Metals, Inc.

2-(3,5-Dimethoxybenzoyl)oxazole

  • MDL number MFCD07699393
  • Cas Number : 898784-40-2
  • Molecular Weight : 233.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-54
Availability details for
Quantity
  • Availability
  • Add to Cart