RIE156322421 Rieke Metals, Inc.

2-(2,3-Dimethylbenzoyl)oxazole

  • MDL number MFCD07699394
  • Cas Number : 898784-42-4
  • Molecular Weight : 201.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5572-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5572-55
Availability details for
Quantity
  • Availability
  • Add to Cart