RIE156322428 Rieke Metals, Inc.

4-(6-Methoxypyridin-3-yl)-4-oxobutyric acid

  • MDL number MFCD07699401
  • Cas Number : 898784-56-0
  • Molecular Weight : 209.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5573B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5573B
Availability details for
Quantity
  • Availability
  • Add to Cart