RIE156322430 Rieke Metals, Inc.

6-(6-Methoxypyridin-3-yl)-6-oxohexanoic acid

  • MDL number MFCD07699403
  • Cas Number : 898784-60-6
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5573D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5573D
Availability details for
Quantity
  • Availability
  • Add to Cart