RIE156322432 Rieke Metals, Inc.

8-(6-Methoxypyridin-3-yl)-8-oxooctanoic acid

  • MDL number MFCD07699405
  • Cas Number : 898784-64-0
  • Molecular Weight : 265.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5573F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5573F
Availability details for
Quantity
  • Availability
  • Add to Cart