RIE156322433 Rieke Metals, Inc.

2-Chloro-4-propionylpyridine

  • MDL number MFCD07699407
  • Cas Number : 87121-53-7
  • Molecular Weight : 169.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-chloro-4-pyridinyl)-1-propanone
Inchi Code 1S/C8H8ClNO/c1-2-7(11)6-3-4-10-8(9)5-6/h3-5H,2H2,1H3
Inchi Key GHONLVKPBHGMJH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5574-01a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-01a
Availability details for
Quantity
  • Availability
  • Add to Cart