RIE156322434 Rieke Metals, Inc.

2-Chloro-4-butyrylpyridine

  • MDL number MFCD07699408
  • Cas Number : 113961-70-9
  • Molecular Weight : 183.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-01B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-01B
Availability details for
Quantity
  • Availability
  • Add to Cart