RIE156322435 Rieke Metals, Inc.

2-Chloro-4-valerylpyridine

  • MDL number MFCD07699409
  • Cas Number : 898784-66-2
  • Molecular Weight : 197.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-01C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-01C
Availability details for
Quantity
  • Availability
  • Add to Cart