RIE156322438 Rieke Metals, Inc.

2-Chloro-4-octanoylpyridine

  • MDL number MFCD07699412
  • Cas Number : 898784-72-0
  • Molecular Weight : 239.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-01F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-01F
Availability details for
Quantity
  • Availability
  • Add to Cart