RIE156322439 Rieke Metals, Inc.

2-Chloro-4-nonanoylpyridine

  • MDL number MFCD07699413
  • Cas Number : 898784-74-2
  • Molecular Weight : 253.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-01G

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-01G
Availability details for
Quantity
  • Availability
  • Add to Cart