RIE156322442 Rieke Metals, Inc.

2-Chloro-4-dodecanoylpyridine

  • MDL number MFCD07699416
  • Cas Number : 898784-80-0
  • Molecular Weight : 295.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-01J

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-01J
Availability details for
Quantity
  • Availability
  • Add to Cart