RIE156322447 Rieke Metals, Inc.

Hexyl 2-chloroisonicotinate

  • MDL number MFCD07699421
  • Cas Number : 898784-90-2
  • Molecular Weight : 241.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name hexyl 2-chloroisonicotinate
Inchi Code 1S/C12H16ClNO2/c1-2-3-4-5-8-16-12(15)10-6-7-14-11(13)9-10/h6-7,9H,2-5,8H2,1H3
Inchi Key SCLCYDPONNYKSU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5574-02e

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-02e
Availability details for
Quantity
  • Availability
  • Add to Cart