RIE156322456 Rieke Metals, Inc.

3-Chloro-4-pyridylcyclopentyl ketone

  • MDL number MFCD07699433
  • Cas Number : 898785-18-7
  • Molecular Weight : 209.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-06D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-06D
Availability details for
Quantity
  • Availability
  • Add to Cart