RIE156322458 Rieke Metals, Inc.

3-Chloro-4-pyridylcycloheptyl ketone

  • MDL number MFCD07699435
  • Cas Number : 898785-24-5
  • Molecular Weight : 237.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-06F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-06F
Availability details for
Quantity
  • Availability
  • Add to Cart