RIE156322461 Rieke Metals, Inc.

1-(3-Chloro-4-pyridyl)-1,5-hexanedione

  • MDL number MFCD07783641
  • Cas Number : 898785-33-6
  • Molecular Weight : 225.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-07C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-07C
Availability details for
Quantity
  • Availability
  • Add to Cart