RIE156322464 Rieke Metals, Inc.

3-Chloro-4-pyridyl (2-cyclohexyl)ethyl ketone

  • MDL number MFCD07699443
  • Cas Number : 898785-45-0
  • Molecular Weight : 251.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-08B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-08B
Availability details for
Quantity
  • Availability
  • Add to Cart