RIE156322465 Rieke Metals, Inc.

3-Chloro-4-pyridyl (3-cyclohexyl)propyl ketone

  • MDL number MFCD07699444
  • Cas Number : 898785-48-3
  • Molecular Weight : 265.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-08C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-08C
Availability details for
Quantity
  • Availability
  • Add to Cart