RIE156322466 Rieke Metals, Inc.

3-Chloro-4-pyridyl (4-cyclohexyl)butyl ketone

  • MDL number MFCD07699445
  • Cas Number : 898785-51-8
  • Molecular Weight : 279.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-08D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-08D
Availability details for
Quantity
  • Availability
  • Add to Cart