RIE156322468 Rieke Metals, Inc.

3-Chloro-4-pyridyl 2-methylpropyl ketone

  • MDL number MFCD07699448
  • Cas Number : 898785-57-4
  • Molecular Weight : 197.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-12
Availability details for
Quantity
  • Availability
  • Add to Cart