RIE156322470 Rieke Metals, Inc.

3-Chloro-4-pyridyl 1-methylbutyl ketone

  • MDL number MFCD07699451
  • Cas Number : 898785-63-2
  • Molecular Weight : 211.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-15
Availability details for
Quantity
  • Availability
  • Add to Cart