RIE156322472 Rieke Metals, Inc.

3-Chloro-4-pyridyl 3-methylbutyl ketone

  • MDL number MFCD07699453
  • Cas Number : 898785-67-6
  • Molecular Weight : 211.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5574-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-17
Availability details for
Quantity
  • Availability
  • Add to Cart