RIE156322473 Rieke Metals, Inc.

3-Chloro-4-pyridyl 1-ethylpropyl ketone

  • MDL number MFCD07699454
  • Cas Number : 898785-69-8
  • Molecular Weight : 211.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-chloro-4-pyridinyl)-2-ethyl-1-butanone
Inchi Code 1S/C11H14ClNO/c1-3-8(4-2)11(14)9-5-6-13-10(12)7-9/h5-8H,3-4H2,1-2H3
Inchi Key LHHSYDZAPQZLKU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5574-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5574-18
Availability details for
Quantity
  • Availability
  • Add to Cart